Learn More
Medchemexpress LLC HGF Human CHO 100ug
SDP
Catalog No. 5000197257
Encompass_Preferred
Recombinant human hepatocyte growth factor (HGF) expressed in CHO cells, supplied as a tag-free, lyophilized protein for research applications in cell proliferation, migration, and tissue regeneration. The product is full-length (isoform-1 mature sequence) and characterized for purity, activity, and low endotoxin to support in vitro assays and signaling studies.
  • Expressed in CHO cells.
  • Supplied tag free and lyophilized for storage stability.
  • Full-length mature sequence (Q32-S728) covering functional domains.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Endotoxin below 0.2 EU/μg as measured by LAL.
  • High specific activity (ED50 <10 ng/mL in 4MBr5 cells).
  • Formulated in PBS or PBS with 8% trehalose; store at -20°C for long term.
Catalog No. 50-001-97257 Supplier Medchemexpress LLC Supplier No. HYP7121100UG
Recombinant human hepatocyte growth factor (HGF) expressed in CHO cells, supplied as a tag-free, lyophilized protein for research applications in cell proliferation, migration, and tissue regeneration. The product is full-length (isoform-1 mature sequence) and characterized for purity, activity, and low endotoxin to support in vitro assays and signaling studies.
  • Expressed in CHO cells.
  • Supplied tag free and lyophilized for storage stability.
  • Full-length mature sequence (Q32-S728) covering functional domains.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Endotoxin below 0.2 EU/μg as measured by LAL.
  • High specific activity (ED50 <10 ng/mL in 4MBr5 cells).
  • Formulated in PBS or PBS with 8% trehalose; store at -20°C for long term.