Learn More
Medchemexpress LLC HGF Human CHO 50ug
SDP
Catalog No. 5000185385
Encompass_Preferred
Recombinant human hepatocyte growth factor (HGF) expressed in CHO cells and supplied lyophilized for research use. The protein is >95% pure by reducing SDS-PAGE, exhibits validated biological activity in cell-based assays, and is offered in multiple microgram-scale sizes suitable for assay development and functional studies.
  • High purity suitable for biochemical and cell-based assays.
  • Expression in CHO cells preserves mammalian glycosylation.
  • Validated biological activity (ED50 <10 ng/mL in 4MBr5 cells).
  • Available in small, convenient package sizes for assay optimization.
  • Lyophilized formulation for improved stability during storage and transport.
  • Stable when aliquoted and stored at -20°C or -80°C for long-term use.
  • Includes amino acid sequence and molecular weight information.
Catalog No. 50-001-85385 Supplier Medchemexpress LLC Supplier No. HYP712150UG
Recombinant human hepatocyte growth factor (HGF) expressed in CHO cells and supplied lyophilized for research use. The protein is >95% pure by reducing SDS-PAGE, exhibits validated biological activity in cell-based assays, and is offered in multiple microgram-scale sizes suitable for assay development and functional studies.
  • High purity suitable for biochemical and cell-based assays.
  • Expression in CHO cells preserves mammalian glycosylation.
  • Validated biological activity (ED50 <10 ng/mL in 4MBr5 cells).
  • Available in small, convenient package sizes for assay optimization.
  • Lyophilized formulation for improved stability during storage and transport.
  • Stable when aliquoted and stored at -20°C or -80°C for long-term use.
  • Includes amino acid sequence and molecular weight information.