Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC HGF Human HEK293 50ug
SDP

Catalog No. 5000185559
Encompass_Preferred

HGF is a recombinant human hepatocyte growth factor produced in HEK293 cells with a C-terminal His tag. Supplied as a lyophilized powder with defined formulation options, it provides high purity and consistent activity for in vitro studies of cell signaling, proliferation, migration, and tissue repair.

  • HEK293-expressed human HGF with C-terminal His tag.
  • Greater than 95% purity as determined by reducing SDS-PAGE.
  • Lyophilized powder with multiple formulation options for stability.
  • Predicted molecular mass of approximately 80.5 kDa; glycosylation yields characteristic α and β subunits on SDS-PAGE.
  • Recommended storage at -20 °C; aliquot and store at -80 °C for extended stability.
  • Suitable for cell signaling and functional assays involving the MET pathway.

Catalog No. 50-001-85559 Supplier Medchemexpress LLC Supplier No. HYP7062750UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

HGF is a recombinant human hepatocyte growth factor produced in HEK293 cells with a C-terminal His tag. Supplied as a lyophilized powder with defined formulation options, it provides high purity and consistent activity for in vitro studies of cell signaling, proliferation, migration, and tissue repair.

  • HEK293-expressed human HGF with C-terminal His tag.
  • Greater than 95% purity as determined by reducing SDS-PAGE.
  • Lyophilized powder with multiple formulation options for stability.
  • Predicted molecular mass of approximately 80.5 kDa; glycosylation yields characteristic α and β subunits on SDS-PAGE.
  • Recommended storage at -20 °C; aliquot and store at -80 °C for extended stability.
  • Suitable for cell signaling and functional assays involving the MET pathway.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.