Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human HGF Instant ELISA™ Kit

Catalog No. 50246657
Encompass_Preferred
Change view
Click to view available options
Quantity:
128 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-246-657 128 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-246-657 Supplier Invitrogen™ Supplier No. BMS2069INST
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

The Human Hepatocyte Growth Factor (Hu HGF) ELISA quantitates Hu HGF in human serum, plasma, cell culture supernatants, or other body fluids. The assay will exclusively recognize both natural and recombinant Hu HGF. Principle of the method The Human HGF solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the binding of the second (detector) antibody to the target on a different epitope from the capture antibody. An antibody conjugated with enzyme binds the formed sandwich. After incubation and washing steps to rid the microplate of unbound substances, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Hepatocyte growth factor regulates cell growth, cell motility, and morphogenesis by activating a tyrosine kinase signaling cascade after binding to the proto-oncogenic c-Met receptor. Hepatocyte growth factor is secreted by mesenchymal cells and acts as a multi-functional cytokine on cells of mainly epithelial origin. Its ability to stimulate mitogenesis, cell motility, and matrix invasion gives it a central role in angiogenesis, tumorogenesis, and tissue regeneration. It is secreted as a single inactive polypeptide and is cleaved by serine proteases into a 69-kDa alpha-chain and 34-kDa beta-chain. A disulfide bond between the alpha and beta chains produces the active, heterodimeric molecule. The protein belongs to the plasminogen subfamily of S1 peptidases but has no detectable protease activity. Alternative splicing of this gene produces multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P14210
Assay Range 63-4000 pg/mL
Assay Sensitivity 5.5 pg/mL
Content And Storage Aluminum pouch(es) with a Microplate coated with antibody against human HGF, Biotin-Conjugate (anti-human HGF antibody), Streptavidin-HRP and Sample Diluent, lyophilized
Aluminum pouch(es) with a human HGF Standard curve (coloured)
1 bottle (25 mL) Wash Buffer Concentrate 20x(phosphate-buffered saline with 1% Tween 20)
1 vial (15 mL) Substrate Solution (tetramethyl-benzidine)
1 vial (12 mL) Sample Diluent (Use when an external predilution of the samples is needed)
1 vial (15 mL) Stop Solution (1M Phosphoric acid)
2 Adhesive Films
Conjugate HRP
Immunoassay Kit Format Coated ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Application) ELISA
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias C230052L06Rik; DFNB39; Fibroblast derived tumor cytotoxic factor; fibroblast-derived tumor cytotoxic factor; F-TCF; hepatocyte growth factor; Hepatocyte growth factor (hepapoietin A; hepatocyte growth factor (hepapoietin A; scatter factor); Hepatocyte growth factor alpha chain; Hepatocyte growth factor beta chain; Hepatopoeitin A; hepatopoeitin-A; HepatopoietinA; hepatopoietin-A; Hgf; HGF/SF; HGFB; HPTA; Lung fibroblast derived mitogen; lung fibroblast-derived mitogen; NK1; NK2; Scatter factor; scatter factor); SF; SF/HGFC230052L06Rik; DFNB39; Fibroblast derived tumor cytotoxic factor; fibroblast-derived tumor cytotoxic factor; F-TCF; hepatocyte growth factor; Hepatocyte growth factor (hepapoietin A; hepatocyte growth factor (hepapoietin A; scatter factor); Hepatocyte growth factor alpha chain; Hepatocyte growth factor beta chain; Hepatopoeitin A; hepatopoeitin-A; HepatopoietinA; hepatopoietin-A; Hgf; HGF/SF; HGFB; HPTA; Lung fibroblast derived mitogen; lung fibroblast-derived mitogen; NK1; NK2; Scatter factor; scatter factor); SF; SF/HGF; DFNB39; F-TCFB; HPTA; SF; HGF
Gene ID (Entrez) 3082
Gene Symbol Hgf
Interassay CV 8.3%
Intraassay CV 7.6%
Kit Contents Pre-coated 96 well Microplate, Biotinylated Detection Antibody, SAV-HRP, Sample Diluent, lyophilized, Standard (8-well strips x 4), Wash Buffer Concentrate, Substrate Solution, Sample Diluent, Stop Solution, Lysis Buffer 10x, Adhesive Plate Covers, Biotinylated Detection Antibody, SAV-HRP, Sample Diluent, lyophilized, Standard (8-well strips x 4), Wash Buffer Concentrate, Substrate Solution, Sample Diluent, Stop Solution, Lysis Buffer 10x, Adhesive Plate Covers
Label or Dye Biotin
Protein Aliases Hepatopoietin-A, Scatter factor, SF
Quantity 128 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements -20°C
Synonym Hepatocyte Growth Factor, hepapoietin A, F-TCF
Target HGF
Test Time 15 min
Total Assay Time 3 hr 10 min
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.