Learn More
AdipoGen HGF human rec
SDP
Supplier:  AdipoGen AG40T0042C025
Encompass_Preferred
Protein. Human HGF 692aa. Source Sf9 cells. Lyophilized. Purity >90% SDS-PAGE. Human hepatocyte growth factor HGF is synthesized as a biologically inactive single chain precursor, which is cleaved by a specific, extracellular serum serine protease to a fully active heterodimer. It stimulates the motility and invasion of several cancer cell types and can induce angiogenesis. All biological responses induced by HGF are elicited by binding to its transmembrane tyrosine kinase receptor. HGF function is essential for normal development. Knockout studies have demonstrated that both ligand and receptor deficient mice display an embryonic lethal phenotype. HGF synergizes with basic FGF in the induction of angiogenesis. HGF is a potent mitogen for mature parenchymal hepatocyte cells and is a pleiotrophic cytokine. It acts as growth factor for a broad spectrum of tissues and cell types. It has no detectable protease activity.
Catalog No. 50-256-2787