Learn More
Medchemexpress LLC HGF Mouse HEK293 100ug
SDP
Catalog No. 5000179107
Encompass_Preferred
HGF Protein a potent mitogen stimulates mature hepatocyte cell growth and acts as a hepatotrophic factor for various tissues As an activating ligand for MET receptor tyrosine kinase HGF binds and promotes dimerization activating MAPK signaling through TMPRSS13 cleavage Structurally HGF comprises alpha and beta chains connected by a disulfide bond Interaction with SRPX2 enhances its mitogenic activity HGF Protein Mouse (HEK293) is the recombinant mouse-derived HGF protein expressed by HEK293 with tag free
Catalog No. 50-001-79107 Supplier Medchemexpress LLC Supplier No. HYP73928100UG
HGF Protein a potent mitogen stimulates mature hepatocyte cell growth and acts as a hepatotrophic factor for various tissues As an activating ligand for MET receptor tyrosine kinase HGF binds and promotes dimerization activating MAPK signaling through TMPRSS13 cleavage Structurally HGF comprises alpha and beta chains connected by a disulfide bond Interaction with SRPX2 enhances its mitogenic activity HGF Protein Mouse (HEK293) is the recombinant mouse-derived HGF protein expressed by HEK293 with tag free