Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC HGF protein, canine (HEK293) | >95.0% | 50 UG
SDP

Catalog No. 5000293134
Encompass_Preferred

Recombinant canine hepatocyte growth factor (HGF) expressed in HEK293 cells, supplied lyophilized and tag-free. HGF binds the MET receptor to promote hepatocyte proliferation, migration, and tissue repair; intended for research use in cell signaling and receptor activation studies. Unit size 50 μg; store frozen and aliquot for extended storage.

  • Suitable for receptor activation and signaling studies.
  • Mammalian-expressed in HEK293 for native-like glycosylation.
  • Tag free to avoid interference in binding assays.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Lyophilized formulation; reconstitute to ≥100 μg/mL and aliquot for storage.

Catalog No. 50-002-93134 Supplier Medchemexpress LLC Supplier No. HYP7488950UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant canine hepatocyte growth factor (HGF) expressed in HEK293 cells, supplied lyophilized and tag-free. HGF binds the MET receptor to promote hepatocyte proliferation, migration, and tissue repair; intended for research use in cell signaling and receptor activation studies. Unit size 50 μg; store frozen and aliquot for extended storage.

  • Suitable for receptor activation and signaling studies.
  • Mammalian-expressed in HEK293 for native-like glycosylation.
  • Tag free to avoid interference in binding assays.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Lyophilized formulation; reconstitute to ≥100 μg/mL and aliquot for storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.