Learn More
Genscript Corporation HGF R/c-METBiotin His Avi
Supplier:  Genscript Corporation Z04478100
Encompass
et also called tyrosine-protein kinase Met or hepatocyte growth factor receptor (HGF R) is a protein that in humans is encoded by the MET gene The protein possesses tyrosine kinase activity The primary single chain precursor protein is post-translationally cleaved to produce the alpha and beta subunits which are disulfide linked to form the mature receptor Following activation by ligand interacts with the PI3-kinase subunit PIK3R1 PLCG1 SRC GRB2 STAT3 or the adapter GAB1
Catalog No. 50-000-59297