Learn More
Medchemexpress LLC HGF Rat HEK293 100ug
SDP
Catalog No. 5000180437
Encompass_Preferred
The HGF gene encodes hepatocyte growth factor (HGF) a protein involved in organ regeneration angiogenesis and myelination It is located in the extracellular space and serves as a biomarker for cholangiocarcinoma Implicated in various diseases HGF plays a crucial role in physiological and pathological conditions It shows biased expression in the liver (RPKM 137 5) lung (RPKM 132 1) and other tissues HGF Protein Rat (HEK293) is the recombinant rat-derived HGF protein expressed by HEK293 with tag free
Catalog No. 50-001-80437 Supplier Medchemexpress LLC Supplier No. HYP73101100UG
The HGF gene encodes hepatocyte growth factor (HGF) a protein involved in organ regeneration angiogenesis and myelination It is located in the extracellular space and serves as a biomarker for cholangiocarcinoma Implicated in various diseases HGF plays a crucial role in physiological and pathological conditions It shows biased expression in the liver (RPKM 137 5) lung (RPKM 132 1) and other tissues HGF Protein Rat (HEK293) is the recombinant rat-derived HGF protein expressed by HEK293 with tag free