Learn More
Enzo Life Sciences HGF SF monoclonal antibody (7-2) (100µg)
SDP
Supplier:  Enzo Life Sciences ADI905165100
Encompass_Preferred
Clone: 7-2. Host: Mouse. Isotype: IgG2a. Immunogen: Human Hepatocyte Growth Factor/Scatter Factor. Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Human, Mouse. Applications: IHC (PS), WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS containing 0.1% sodium azide.
Catalog No. 50-200-6938