Learn More
Medchemexpress LLC HGFA/HGF activator protein, human | >95.0% | 10 UG
SDP
Catalog No. 5000290947
Encompass_Preferred
HGFA/HGF Activator Protein, Human (HEK293, His) is a recombinant human hepatocyte growth factor activator expressed in HEK293 cells with a C-terminal His tag. It catalyzes conversion of pro-HGF into its active heterodimer and is supplied lyophilized for research applications.
  • Human recombinant protein expressed in HEK293 cells.
  • C-terminal His tag for detection and purification.
  • Lyophilized formulation from 0.22 μm filtered PBS, pH 7.4, with 5-8% trehalose.
  • Purity greater than 95% as determined by reducing SDS-PAGE (measured 96.9%).
  • Apparent molecular weight approximately 95-110 kDa.
  • Suitable for research applications involving HGF activation and signaling.
Catalog No. 50-002-90947 Supplier Medchemexpress LLC Supplier No. HYP7638010UG
HGFA/HGF Activator Protein, Human (HEK293, His) is a recombinant human hepatocyte growth factor activator expressed in HEK293 cells with a C-terminal His tag. It catalyzes conversion of pro-HGF into its active heterodimer and is supplied lyophilized for research applications.
  • Human recombinant protein expressed in HEK293 cells.
  • C-terminal His tag for detection and purification.
  • Lyophilized formulation from 0.22 μm filtered PBS, pH 7.4, with 5-8% trehalose.
  • Purity greater than 95% as determined by reducing SDS-PAGE (measured 96.9%).
  • Apparent molecular weight approximately 95-110 kDa.
  • Suitable for research applications involving HGF activation and signaling.