Learn More
Sino Biological HGF/Hepatocyte Growth Factor Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Catalog No. 502248221
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human HGF/Hepatocyte Growth Factor (rh HGF/Hepatocyte Growth Factor; Catalog#10463-HNAS; NP_000592.3; Met1-Ser728). HGF/Hepatocyte Growth Factor specific IgG was purified by Human HGF/Hepatocyte Growth Factor affinity chromatography.
This item is not returnable. View return policy
Catalog No. 50-224-8221 Supplier Sino Biological Supplier No. 10463T24100
Produced in rabbits immunized with purified, recombinant Human HGF/Hepatocyte Growth Factor (rh HGF/Hepatocyte Growth Factor; Catalog#10463-HNAS; NP_000592.3; Met1-Ser728). HGF/Hepatocyte Growth Factor specific IgG was purified by Human HGF/Hepatocyte Growth Factor affinity chromatography.