Learn More
Cayman Chemical HGFhumnrecombInt 100ug
SDP
Catalog No. 502947415
Encompass_Preferred
Source: Active recombinant human HGF expressed in CHO cells • Amino acids: 32-728 • MW: 79.7 kDa
Catalog No. 50-294-7415 Supplier Cayman Chemical Supplier No. 32052100 UG
Source: Active recombinant human HGF expressed in CHO cells • Amino acids: 32-728 • MW: 79.7 kDa