Learn More
ACROBiosystems ACROBiosystems Biotinylated Human HGF R / c-MET Protein, Avitag™,His Tag
Catalog No. 501957428
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human HGF R, Avitag,His Tag (MET-H82E1) is expressed from human 293 cells (HEK293). It contains AA Glu 25 - Thr 932 (Accession # AAI30421.1).;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 104.3 kDa. The protein migrates as 40-45 kDa α subunit and 80 kDa β subunit under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human HGF protein at 10 μg/mL (100 μL/well) can bind Biotinylated Human HGF R, Avitag,His Tag (Cat. No. MET-H82E1) with a linear range of 0.04-0.31 μg/mL (QC tested).
Catalog No. 50-195-7428 Supplier ACROBiosystems Supplier No. METH82E125UG
Recombinant Protein;25UG;Biotinylated Human HGF R, Avitag,His Tag (MET-H82E1) is expressed from human 293 cells (HEK293). It contains AA Glu 25 - Thr 932 (Accession # AAI30421.1).;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 104.3 kDa. The protein migrates as 40-45 kDa α subunit and 80 kDa β subunit under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human HGF protein at 10 μg/mL (100 μL/well) can bind Biotinylated Human HGF R, Avitag,His Tag (Cat. No. MET-H82E1) with a linear range of 0.04-0.31 μg/mL (QC tested).