Learn More
ACROBiosystems Recombinant Protein;Human HGF R / c-MET Protein, Fc Tag;HEK293;1MG;MET,MET,AUTS9,HGFR,RCCP2,c-Met
Supplier:  ACROBiosystems METH52561MG
Encompass_Preferred
Recombinant Protein;Human HGF R / c-MET Protein, Fc Tag;1MG;Human HGF R, Fc Tag (MET-H5256) is expressed from human 293 cells (HEK293). It contains AA Glu 25 - Thr 932 (Accession # AAI30421.1).;rhHGFR is fused with Fc fragment of human IgG1 at the C-terminus. The mature form of HGFR is a disulfide-linked heterodimer composed of proteolytically cleaved α and β chain. Each α and β chain has a calculated MW of 32.5 kDa (α chain) and 96.7 kDa (β chain Fc chimera). The predicted N-terminal is Glu25 (α chain) & Ser308 (β chain Fc chimera). Protein migrates as 45 kDa (α chain) and 120-125 kDa (βchain Fc chimera) in reduced SDS-PAGE resulting from glycosylation.
Catalog No. 50-165-7966