Learn More
ACROBiosystems Recombinant Protein;Human HGF R / c-MET Protein;HEK293;200UG;MET,MET,AUTS9,HGFR,RCCP2,c-Met
Supplier:  ACROBiosystems METH5227200UG
Encompass_Preferred
Recombinant Protein;Human HGF R / c-MET Protein;200UG;Human HGF R, His Tag (MET-H5227) is expressed from human 293 cells (HEK293). It contains AA Glu 25 - Thr 932 (Accession # AAI30421.1).;rhHGFR is fused with a polyhistidine tag at the C-terminal. The mature form of HGFR is a disulfide-linked heterodimer composed of proteolytically cleaved α and β chain. Each α and β chain has a calculated MW of 32.5 kDa (α chain) and 60 kDa (β chain). The predicted N-terminal is Glu25 (α chain) & Ser308 (β chain). Protein migrates as 45 kDa (α chain) and 85-90 kDa (β chain) in reduced SDS-PAGE resulting from glycosylation. ;Immobilized Human HGF/Hepatocyte Growth Factor at 5μg/mL (100 μL/well) can bind Human HGF R, His Tag (Cat. No. MET-H5227) with a linear range of 5-78 ng/mL (Routinely tested).
Catalog No. 50-165-7617