Learn More
Electron Microscopy Sciences High Precision Cutters, Head: Micro Oval. OAL: 120 mm., EA
SDP
Catalog No. NC1881560
Encompass_Preferred
High precision Cutters. ESD safe Medical Device Cutters, Full-flush - open springs. Head: Micro Oval. OAL: 120 mm .
Catalog No. NC1881560 Supplier Electron Microscopy Sciences Supplier No. 9520
High precision Cutters. ESD safe Medical Device Cutters, Full-flush - open springs. Head: Micro Oval. OAL: 120 mm .