Learn More
Magbio Genomics Inc HighPrep Blood & Tissue DNA Kit (96 preps)
SDP
Catalog No. 501656594
Encompass_Preferred
The HighPrep™ Blood & Tissue DNA kit is a magnetic beads based system to process high quality genomic DNA from 50-250 µL of fresh or frozen whole blood, buffy coat (containing anticoagulants such as Citrate, EDTA and Heparin), lysate of tissues, mouse tails, cultured cells, saliva, or buccal swabs.The HighPrep Blood & Tissue DNA kit is highly geared for automation where high quality genomic DNA is reproducible in a high throughput environment. The kit is adaptable to commercially available liquid handling workstations.
Catalog No. 50-165-6594 Supplier Magbio Genomics Inc Supplier No. HPBTSD96
The HighPrep™ Blood & Tissue DNA kit is a magnetic beads based system to process high quality genomic DNA from 50-250 µL of fresh or frozen whole blood, buffy coat (containing anticoagulants such as Citrate, EDTA and Heparin), lysate of tissues, mouse tails, cultured cells, saliva, or buccal swabs.The HighPrep Blood & Tissue DNA kit is highly geared for automation where high quality genomic DNA is reproducible in a high throughput environment. The kit is adaptable to commercially available liquid handling workstations.