Learn More
Magbio Genomics Inc HighPrep Plasmid DNA Kit (96 preps)
SDP
Catalog No. 501656596
Encompass_Preferred
Magbio's HighPrep™ Plasmid DNA kit is a magnetic beads based plasmid purification system for high throughput rapid isolation of high and low copy number plasmid DNA, BACs, PACs Cosmid and Fosmids. The system can be easily automated on many liquid handling workstation providing robustness and high quality plasmid DNA for subsequent downstream applications such as digestion and fluorescent DNA sequencing
Catalog No. 50-165-6596 Supplier Magbio Genomics Inc Supplier No. HPPLDP96
Magbio's HighPrep™ Plasmid DNA kit is a magnetic beads based plasmid purification system for high throughput rapid isolation of high and low copy number plasmid DNA, BACs, PACs Cosmid and Fosmids. The system can be easily automated on many liquid handling workstation providing robustness and high quality plasmid DNA for subsequent downstream applications such as digestion and fluorescent DNA sequencing