Learn More
Magbio Genomics Inc HighPrep Plasmid DNA Kit (384 preps)
SDP
Catalog No. 501656597
Encompass_Preferred
Magbio's HighPrep™ Plasmid DNA kit is a magnetic beads based plasmid purification system for high throughput rapid isolation of high and low copy number plasmid DNA, BACs, PACs Cosmid and Fosmids. The system can be easily automated on many liquid handling workstation providing robustness and high quality plasmid DNA for subsequent downstream applications such as digestion and fluorescent DNA sequencing
Catalog No. 50-165-6597 Supplier Magbio Genomics Inc Supplier No. HPPLDP96X4
Magbio's HighPrep™ Plasmid DNA kit is a magnetic beads based plasmid purification system for high throughput rapid isolation of high and low copy number plasmid DNA, BACs, PACs Cosmid and Fosmids. The system can be easily automated on many liquid handling workstation providing robustness and high quality plasmid DNA for subsequent downstream applications such as digestion and fluorescent DNA sequencing