Learn More
Cayman Chemical H-Gly-Pro-Arg-Pro-OH (acetate), Purity: ≥95%, CAS: 157009-81-9, 10 mg
SDP
Catalog No. 502983466
Encompass_Preferred
A peptide corresponding partly to the amino-terminus of the Aα chain and Bβ chain of fibrin; inhibits fibrin polymerization and ADP-induced platelet aggregation (IC50 = 3 mM)
Catalog No. 50-298-3466 Supplier Cayman Chemical Supplier No. 2116310 MG
A peptide corresponding partly to the amino-terminus of the Aα chain and Bβ chain of fibrin; inhibits fibrin polymerization and ADP-induced platelet aggregation (IC50 = 3 mM)