Learn More
Medchemexpress LLC Hh-Ag1.5 (SAg1.5) | 612542-14-0 | ≥98.0% | 526.04 | C28H26ClF2N3OS | 10MG
SDP
Catalog No. 5000239067
Encompass_Preferred
Hh-Ag1.5 is a small-molecule agonist of the Hedgehog signaling pathway that activates Smoothened (Smo) and induces Gli-dependent transcription. It is used in vitro to stimulate Hh signaling, with reported activity at subnanomolar concentrations and documented application in reprogramming and expansion of quiescent liver stem cells.
  • Potent Hedgehog pathway agonist with EC50 ≈ 1 nM.
  • Selective Smoothened (Smo) receptor agonist with Ki ≈ 0.52 nM.
  • Facilitates expansion of quiescent liver stem cells in vitro at 5 μM.
  • High purity suitable for cellular assays (≥98% by HPLC).
  • Molecular weight 526.04; formula C28H26ClF2N3OS; soluble in DMSO.
Catalog No. 50-002-39067 Supplier Medchemexpress LLC Supplier No. HY12489910MG
Hh-Ag1.5 is a small-molecule agonist of the Hedgehog signaling pathway that activates Smoothened (Smo) and induces Gli-dependent transcription. It is used in vitro to stimulate Hh signaling, with reported activity at subnanomolar concentrations and documented application in reprogramming and expansion of quiescent liver stem cells.
  • Potent Hedgehog pathway agonist with EC50 ≈ 1 nM.
  • Selective Smoothened (Smo) receptor agonist with Ki ≈ 0.52 nM.
  • Facilitates expansion of quiescent liver stem cells in vitro at 5 μM.
  • High purity suitable for cellular assays (≥98% by HPLC).
  • Molecular weight 526.04; formula C28H26ClF2N3OS; soluble in DMSO.