Learn More
New England Biolabs, Inc. HhaI Methyltransferase – 1000 units
SDP
Catalog No. 50811636
Encompass
HhaI Methyltransferase modifies the internal cytosine residue (C5) of the sequence GCGC
Catalog No. 50-811-636 Supplier New England Biolabs, Inc. Supplier No. M0217S
HhaI Methyltransferase modifies the internal cytosine residue (C5) of the sequence GCGC