Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex™ HHAAD Headgear Adapter

Catalog No. 19155362 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Material:
Aluminum Alloy
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-155-362 Aluminum Alloy
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-155-362 Supplier Pyramex™ Supplier No. HHAAD
Add to Cart
Add to Cart

Deluxe aluminum cap style hard hat adapter

  • Constructed from light weight aluminum
  • Designed to fit more rounded cap style hard hats
  • Accommodates a variety of face shields - does not fit S1060
  • Specifications

    Certifications/Compliance ANSI Z87.1
    Color Silver
    For Use With (Equipment) Rounded, Cap-style Hard Hats
    Material Aluminum Alloy
    Thickness 1.0 to 1.45 mm
    Weight (English) 5.6 oz.
    Weight (Metric) 158 g
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.