Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex™ HHAAW Headgear Adapter

Catalog No. 19155363 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Material:
Aluminum Alloy
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-155-363 Aluminum Alloy
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-155-363 Supplier Pyramex™ Supplier No. HHAAW
Add to Cart
Add to Cart
This item is not returnable. View return policy

Aluminum full brim hard hat adapter

  • Constructed from light weight aluminum
  • Fits most full brim hard hats and accommodates a wide variety of face shields
  • Specifications

    Certifications/Compliance ANSI Z87.1
    Color Silver
    For Use With (Equipment) Fits most full brim hard hats and accommodates a wide variety of face shields
    Material Aluminum Alloy
    Thickness 1.0 to 1.45 mm
    Weight (English) 5.6 oz.
    Weight (Metric) 158 g
    Head Style Wide Brim
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.