Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex™ HHABCMS Headgear Adapter

Catalog No. 19155366 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Material:
Nylon, ABS
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-155-366 Nylon, ABS
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-155-366 Supplier Pyramex™ Supplier No. HHABCMS
Add to Cart
Add to Cart

SL Series Cap Style hard hat adapter

  • Dielectric
  • Hard hat adapters fit into slots of cap style hard hats
  • Can be used with cap mounted earmuff
  • Adjustable while using cap mounted earmuff
  • Fits SL Series cap style hard hats and accommodates a variety of face shields
  • Specifications

    Certifications/Compliance ANSI Z87.1
    Color Black
    For Use With (Equipment) Rounded, Cap-style Hard Hats
    Material Nylon, ABS
    Thickness 2.1 to 2.9 mm
    Weight (English) 3.17 oz.
    Weight (Metric) 90 g
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.