Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex™ HHABW Headgear Adapter

Catalog No. 19155367 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Material:
ABS
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-155-367 ABS
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-155-367 Supplier Pyramex™ Supplier No. HHABW
Add to Cart
Add to Cart

Dielectric full brim hard hat adapter

  • Dielectric
  • Constructed from High Impact Resistant ABS Resin
  • Fits on most full brim hard hats and accommodates a variety of face shields
  • Specifications

    Certifications/Compliance ANSI Z87.1
    Color Black
    For Use With (Equipment) Full Brim Hard Hats
    Material ABS
    Weight (English) 3.1 oz.
    Weight (Metric) 88 g
    Head Style Wide Brim
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.