Learn More
Aat Bioquest HHBS [Hanks Buffer with 20 mM Hepes] (100 mL)
SDP
Catalog No. 502249795
Encompass_Preferred
Hanks Balanced Salt Solution with 20 mM Hepes buffer HHBS is used for a variety of cell culture applications such as washing cells transporting cells or tissue samples diluting cells for counting and preparing reagents. This catalog is for 100 mL.
This item is not returnable. View return policy
Catalog No. 50-224-9795 Supplier Aat Bioquest Supplier No. 20011
Hanks Balanced Salt Solution with 20 mM Hepes buffer HHBS is used for a variety of cell culture applications such as washing cells transporting cells or tissue samples diluting cells for counting and preparing reagents. This catalog is for 100 mL.