Learn More
Aviva Systems Biology Corp Hhex Antibody - C-terminal region (ARP36879_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36879_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Hhex. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the c terminal region of mouse Hhex. Alternate names: Hex, Hhex-rs2, Prh, PrhxApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-62931