Learn More
Aviva Systems Biology Corp HHEX Antibody - middle region (ARP31443_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31443_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HHEX. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human HHEX. Alternate names: HEX, PRH, HMPH, PRHX, HOX11L-PENApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-54225