Learn More
Aviva Systems Biology Corp HHIPL1 Antibody - C-terminal region (ARP59843_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59843_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HHIPL1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human HHIPL1. Alternate names: KIAA1822, UNQ9245ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-286-850