Learn More
iXCells Biotechnologies Human Hepatic Stellate Cells- Adult (HHSC)
Catalog No. 501078058
Encompass_Preferred
iXCells Biotechnologies provides high quality Human Hepatic Stellate Cells (HHSC), which are isolated from adult human liver and cryopreserved with >1 million cells in each vial. Each vial is sufficient to seed on four 10 cm dishes, and cells will be ready in 5~6 days to be passaged at 1:3 ratio. HUSMC express Desmin and GFAP, and are negative for HIV-1, HBV, HCV, mycoplasma, bacteria, yeast, and fung. HUSMC can further expand for 2-3 population doublings in¿Stellate Cell Growth Medium¿under the condition suggested by iXCells Biotechnologies.
Catalog No. 50-107-8058 Supplier iXCells Biotechnologies Supplier No. 10HU210
iXCells Biotechnologies provides high quality Human Hepatic Stellate Cells (HHSC), which are isolated from adult human liver and cryopreserved with >1 million cells in each vial. Each vial is sufficient to seed on four 10 cm dishes, and cells will be ready in 5~6 days to be passaged at 1:3 ratio. HUSMC express Desmin and GFAP, and are negative for HIV-1, HBV, HCV, mycoplasma, bacteria, yeast, and fung. HUSMC can further expand for 2-3 population doublings in¿Stellate Cell Growth Medium¿under the condition suggested by iXCells Biotechnologies.