Learn More
iXCells Biotechnologies Human Hepatic Sinusoidal Endothelial Cells (HHSEC, Adult)
Catalog No. 501078013
Encompass_Preferred
iXCells Biotechnologies provides high quality HHSEC, which are isolated from normal human liver and cryopreserved at P2, with >0.5 million cells in each vial. HHSEC express vWF/Factor VIII and CD31. They are negative for HIV-1, HBV, HCV, mycoplasma, bacteria, yeast, and fungi and can further expand in Endothelial Cell Growth Medium under the condition suggested by iXCells Biotechnologies.
Catalog No. 50-107-8013 Supplier iXCells Biotechnologies Supplier No. 10HU021
iXCells Biotechnologies provides high quality HHSEC, which are isolated from normal human liver and cryopreserved at P2, with >0.5 million cells in each vial. HHSEC express vWF/Factor VIII and CD31. They are negative for HIV-1, HBV, HCV, mycoplasma, bacteria, yeast, and fungi and can further expand in Endothelial Cell Growth Medium under the condition suggested by iXCells Biotechnologies.