Learn More
ACROBiosystems HHV-5 (strain AD169) Glycoprotein H&L, His Tag (MALS verified)
Catalog No. 502719594
Encompass_Preferred
Human cytomegalovirus (HCMV), a prototypical beta-herpes virus, is a major cause of morbidity and mortality in immunocompromised transplant recipients and congenitally infected fetuses.Different HCMV glycoprotein complexes are capable of meditating entry through interaction with cell-specific receptors on a panel of host cells. The pentamer (gH/gL/UL128/UL130/UL131A) mediates HCMV entry into epithelial, endothelial, and myeloid cells by its binding to neuropilin 2 (Nrp2). The trimer (gH/gL/gO) can mediate infection of all cell types by binding to platelet-derived growth factor-alpha (PDGFRα). Both pentamer and trimer need to interact with the glycoprotein B (gB), the fusogenic protein, to trigger the virus and host cell membrane fusion.
Catalog No. 50-271-9594 Supplier ACROBiosystems Supplier No. GHLH5243500UG
Human cytomegalovirus (HCMV), a prototypical beta-herpes virus, is a major cause of morbidity and mortality in immunocompromised transplant recipients and congenitally infected fetuses.Different HCMV glycoprotein complexes are capable of meditating entry through interaction with cell-specific receptors on a panel of host cells. The pentamer (gH/gL/UL128/UL130/UL131A) mediates HCMV entry into epithelial, endothelial, and myeloid cells by its binding to neuropilin 2 (Nrp2). The trimer (gH/gL/gO) can mediate infection of all cell types by binding to platelet-derived growth factor-alpha (PDGFRα). Both pentamer and trimer need to interact with the glycoprotein B (gB), the fusogenic protein, to trigger the virus and host cell membrane fusion.