Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ HHV-8 ORF K4 Recombinant Protein, PeproTech®

Catalog No. 35003250UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
250 μg
50 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
35003250UG 250 μg
35003100UG 100 μg
350-03-10UG 10 μg
350-03-1MG 1 mg
35003500UG 500 μg
350-03-50UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 35003250UG Supplier Gibco™ Supplier No. 35003250UG
  • $602.00
    $451.65 / Each of 1

    Save $150.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Recombinant Protein

Recombinant Viral MIP-2 is a 7.9 kDa protein consisting of 70 amino acids, including the four highly conserved cysteine residues present in CC chemokines. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Viral MIP-2 (vMIP-2) is a chemokine analog encoded by the human herpes virus, and has been shown to have antagonist activity towards several chemokine receptors.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q98157
For Use With (Application) Bioactivity, Control
Formulation Protein with no preservative
Gene ID (Entrez) 4961514
Molecular Weight (g/mol) 7.9 kDa
Name HHV-8 ORF K4
Purification Method Purified
Quantity 250 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Common Name HHV-8 ORF K4
Gene Symbol K4
Biological Activity Determined by the inhibitory effect on monocyte migration response to human MIP-1 alpha using a concentration range of 1.0 ug-10.0 ug/ml of viral MIP-2 will inhibit 25 ng/ml of human MIP-1 alpha.
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence LGASWHRPDK CCLGYQKRPL PQVLLSSWYP TSQLCSKPGV IFLTKRGRQV CADKSKDWVK KLMQQLPVTA
Content And Storage -20°C
Activity Determined by the inhibitory effect on monocyte migration response to human MIP-1 alpha using a concentration range of 1.0ug-10.0ug/ml of viral MIP-2 will inhibit 25ng/ml of human MIP-1 alpha.
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.