Learn More
Diagnostic Biosystems HHV8 HHV8
Catalog No. 502407504
Encompass_Preferred
HHV 8 encodes a latent nuclear antigen (LNA), which is the product of the viral gene orf 73. LNA is capable of forming a complex with retinoblastoma susceptibility gene product, which may be related to its oncogenic activity. HHV8 is associated with three different diseases observed in AIDS patients;  kaposi's sarcoma, primary effusion lymphoma (which is a rare type of nonHodgkin lymphoma affecting the body cavities), and multicentric Castleman's disease. HHV 8 is the likely etiological agent of Kaposi’s sarcoma.
Catalog No. 50-240-7504 Supplier Diagnostic Biosystems Supplier No. PDM395
HHV 8 encodes a latent nuclear antigen (LNA), which is the product of the viral gene orf 73. LNA is capable of forming a complex with retinoblastoma susceptibility gene product, which may be related to its oncogenic activity. HHV8 is associated with three different diseases observed in AIDS patients;  kaposi's sarcoma, primary effusion lymphoma (which is a rare type of nonHodgkin lymphoma affecting the body cavities), and multicentric Castleman's disease. HHV 8 is the likely etiological agent of Kaposi’s sarcoma.