Learn More
MSC HI CAP MNTNC ABSORB PAD
Catalog No. 5000796807
Encompass
These sorbent pads are designed for universal use, effectively handling most non-aggressive liquids. Featuring a highly absorbent, low-profile design, they are ideal for placement in, under, and around machinery.
  • Made from polyester and polypropylene
  • Highly absorbent, low-profile design
  • Perforated
  • Suitable for placement in, under, and around machines
  • Effective on most non-aggressive liquids
Catalog No. 50-007-96807 Supplier MSC Supplier No. 17015405
These sorbent pads are designed for universal use, effectively handling most non-aggressive liquids. Featuring a highly absorbent, low-profile design, they are ideal for placement in, under, and around machinery.
  • Made from polyester and polypropylene
  • Highly absorbent, low-profile design
  • Perforated
  • Suitable for placement in, under, and around machines
  • Effective on most non-aggressive liquids