Learn More
Darwin Chambers Company HI DENSITY LIGHT OPTION
SDP
Catalog No. NC0869750
Encompass_Preferred
High intensity light option. (1) 24" LED lamp, 615 lumens, color 6300K, 7.2 watts. Mounted on the center of door unless specified. Lighting options may change low temperature range specifications.
Catalog No. NC0869750 Supplier Darwin Chambers Company Supplier No. LTG7
High intensity light option. (1) 24" LED lamp, 615 lumens, color 6300K, 7.2 watts. Mounted on the center of door unless specified. Lighting options may change low temperature range specifications.