Learn More
HIMEDIA Hi-FlexiLoop 4 Sterilized Flexifle 0.01mL 5x100 (500/cs)
Catalog No. NC3429567
Encompass_Preferred
Hi-FlexiLoop 4 Sterilized Flexifle 0.01mL 5x100 (500/cs)
Catalog No. NC3429567 Supplier HIMEDIA Supplier No. PW0135X100NO
Hi-FlexiLoop 4 Sterilized Flexifle 0.01mL 5x100 (500/cs)