Learn More
Azzota Corp Ultrasonic Cleaner, Model Bhf200, 53 Khz Frequency, 10L Capacity, 200 W, 1-99 Min. Timer, Lcd Display, With Drain
Catalog No. 501612784
Encompass_Preferred
Ultrasonic Cleaner, Model BHF200, 53 kHz Frequency, 10L Capacity, 200 W, 1-99 Min. Timer, LCD Display, with Drain
Catalog No. 50-161-2784 Supplier Azzota Corp Supplier No. BHF200
Ultrasonic Cleaner, Model BHF200, 53 kHz Frequency, 10L Capacity, 200 W, 1-99 Min. Timer, LCD Display, with Drain