Learn More
Azzota Corp High-Frequency Adjustable Power Ultrasonic Cleaners, Benchtop, 53 Khz, 10 L, 200W, 20C-60C, Indies Size: 34X15X24Cm, 1-199 Min.
Catalog No. 501612780
Encompass_Preferred
High-frequency Adjustable Power Ultrasonic Cleaners, Benchtop, 53 kHz, 10 L, 200W, 20C-60C, Indies size: 34x15x24cm, 1-199 Min.
Catalog No. 50-161-2780 Supplier Azzota Corp Supplier No. BHA200T
High-frequency Adjustable Power Ultrasonic Cleaners, Benchtop, 53 kHz, 10 L, 200W, 20C-60C, Indies size: 34x15x24cm, 1-199 Min.