Learn More
Azzota Corp Ultrasonic Cleaner, Model Bhf-50, 53 Khz Frequency, 2.0L Capacity, 50 W, 1-99 Min. Timer, Lcd Display
Catalog No. 501612785
Encompass_Preferred
Ultrasonic Cleaner, Model BHF-50, 53 kHz Frequency, 2.0L Capacity, 50 W, 1-99 Min. Timer, LCD Display
Catalog No. 50-161-2785 Supplier Azzota Corp Supplier No. BHF50
Ultrasonic Cleaner, Model BHF-50, 53 kHz Frequency, 2.0L Capacity, 50 W, 1-99 Min. Timer, LCD Display