Learn More
Azzota Corp High-Frequency Adjustable Power Ultrasonic Cleaners, Benchtop, 53 Khz, 100W, 3 L, 20C-60C, Indies Size: 24X14X10Cm, 1-199 Min.
Supplier:  Azzota Corp BHA100T
Encompass_Preferred
High-frequency Adjustable Power Ultrasonic Cleaners, Benchtop, 53 kHz, 100W, 3 L, 20C-60C, Indies size: 24x14x10cm, 1-199 Min.
Catalog No. 50-161-2778