Learn More
Azzota Corp Ultrasonic Cleaner, Model Bhf180, 53 Khz Frequency, 6L Capacity, 180 W, 1-99 Min. Timer, Lcd Display
Catalog No. 501612783
Encompass_Preferred
Ultrasonic Cleaner, Model BHF180, 53 kHz Frequency, 6L Capacity, 180 W, 1-99 Min. Timer, LCD Display
Catalog No. 50-161-2783 Supplier Azzota Corp Supplier No. BHF180
Ultrasonic Cleaner, Model BHF180, 53 kHz Frequency, 6L Capacity, 180 W, 1-99 Min. Timer, LCD Display