Learn More
Azzota Corp High-Frequency Adjustable Power Ultrasonic Cleaners, Benchtop, 53 Khz, 6 L, 180W, 20C-60C, Indies Size: 30X15X10Cm, 1-199 Min.
Catalog No. 501612779
Encompass_Preferred
High-frequency Adjustable Power Ultrasonic Cleaners, Benchtop, 53 kHz, 6 L, 180W, 20C-60C, Indies size: 30x15x10cm, 1-199 Min.
Catalog No. 50-161-2779 Supplier Azzota Corp Supplier No. BHA180T
High-frequency Adjustable Power Ultrasonic Cleaners, Benchtop, 53 kHz, 6 L, 180W, 20C-60C, Indies size: 30x15x10cm, 1-199 Min.