Learn More
Electron Microscopy Sciences Hi/Lo Cloth Temperature Tape, 1/2" Wide 40 YD
SDP
Catalog No. 5031293
Encompass_Preferred
These tapes mold tightly to any container, withstands acids, alkalis, oils and greases. It's will not crack, peel, or fall off in 550 degree F dry oven temperatures. Hi/Lo Temperature tape also sticks tightly. This tape must be cut with scissors.
Catalog No. 50-312-93 Supplier Electron Microscopy Sciences Supplier No. 7701205
These tapes mold tightly to any container, withstands acids, alkalis, oils and greases. It's will not crack, peel, or fall off in 550 degree F dry oven temperatures. Hi/Lo Temperature tape also sticks tightly. This tape must be cut with scissors.