Learn More
Sigma Aldrich Fine Chemicals Biosciences Hi/Lo profile rocker Rocke
Catalog No. 502908593
Encompass
The heavy-duty Hi Low Profile Rocker unit is designed for versatility. It operates on bench tops in high humidity or dry incubators in high temperature ovens (to 65C) and refrigerated units (to 0C). Typical applications include Staining and Destaining Gels Northern and Southern Hybridizations Western Blotting and Rinsing Membranes Cell Culture Incubation and Phage Elutions or Amplification of cDNA libraries.The Hi Lo Profile Rocker fits easily into laboratory incubators ovens and refrigerated units without rearranging shelves or disturbing other materials. The thin flat power cord reaches external outlets without damaging incubator or refrigerator gaskets. The platform is continuously adjustable from 2 to 11 and can be effortlessly changed in 15 seconds. The speed varies from 0 - 22rpm with no cogging at slow speeds.All finishes and hardware are stainless steel and the rocker can be easily disassembled for sterilizing in the autoclave. The rocker can hold a maximum load of 2
Catalog No. 50-290-8593 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7421991KT
The heavy-duty Hi Low Profile Rocker unit is designed for versatility. It operates on bench tops in high humidity or dry incubators in high temperature ovens (to 65C) and refrigerated units (to 0C). Typical applications include Staining and Destaining Gels Northern and Southern Hybridizations Western Blotting and Rinsing Membranes Cell Culture Incubation and Phage Elutions or Amplification of cDNA libraries.The Hi Lo Profile Rocker fits easily into laboratory incubators ovens and refrigerated units without rearranging shelves or disturbing other materials. The thin flat power cord reaches external outlets without damaging incubator or refrigerator gaskets. The platform is continuously adjustable from 2 to 11 and can be effortlessly changed in 15 seconds. The speed varies from 0 - 22rpm with no cogging at slow speeds.All finishes and hardware are stainless steel and the rocker can be easily disassembled for sterilizing in the autoclave. The rocker can hold a maximum load of 2