Learn More
Electron Microscopy Sciences Hi/Lo Write-On™ Temperature Tape ½", White, 3" Core 40 YD
SDP
Catalog No. 5029949
Encompass_Preferred
These color-coded label tapes work well with all glass and plastic labware, racks, tubing, bags, boxes, and cabinets. It adheres to any surface and peels off without leaving a sticky residue. Tapes resist moisture and most solvents and can withstand autoclaving (250 degree F). It may be used as well down to -73 degree C.
Width: ½"
Color: White
Core: 3"
Catalog No. 50-299-49 Supplier Electron Microscopy Sciences Supplier No. 77010W1
These color-coded label tapes work well with all glass and plastic labware, racks, tubing, bags, boxes, and cabinets. It adheres to any surface and peels off without leaving a sticky residue. Tapes resist moisture and most solvents and can withstand autoclaving (250 degree F). It may be used as well down to -73 degree C.
Width: ½"
Color: White
Core: 3"