Learn More
MSC ZEP Floor Cleaner, Liquid, 1 Gal Bottle, Grease Digester, 4/Pk
Catalog No. 502817408
Encompass
ZEP Bio Swab Floor Cleaner Grease Digester is a liquid, green floor care product designed for use on various hard and soft floor types, including quarry tile, concrete, ceramic, terrazzo, vinyl, and rubber. This cleaner is formulated to effectively digest grease.
  • Liquid form
  • Green color
  • Characteristic odor
  • Has a pH range of 8 to 9
  • Nonflammable
  • Soluble in cold water
  • EcoLogo certified
  • Container size of 1 gal
  • Packaged as 4 bottles
  • Recommended dilution 1:32 to 1:128
  • Maximum coverage area 16,000 sq ft per gallon
Catalog No. 50-281-7408 Supplier MSC Supplier No. 17690397
ZEP Bio Swab Floor Cleaner Grease Digester is a liquid, green floor care product designed for use on various hard and soft floor types, including quarry tile, concrete, ceramic, terrazzo, vinyl, and rubber. This cleaner is formulated to effectively digest grease.
  • Liquid form
  • Green color
  • Characteristic odor
  • Has a pH range of 8 to 9
  • Nonflammable
  • Soluble in cold water
  • EcoLogo certified
  • Container size of 1 gal
  • Packaged as 4 bottles
  • Recommended dilution 1:32 to 1:128
  • Maximum coverage area 16,000 sq ft per gallon