Learn More
Cowie Technology Corporation HI PERFORMANCE SHAFT GUIDE
Catalog No. NC2207783
Encompass_Preferred
For use with Glass and Metal Stirrer Shafts, Shaft diameter: 8.0 mm, Cone:24/40, Height: 60 mm, Diameter: 42 mm
This item is not returnable. View return policy
Catalog No. NC2207783 Supplier Cowie Technology Corporation Supplier No. 005.0.0824
For use with Glass and Metal Stirrer Shafts, Shaft diameter: 8.0 mm, Cone:24/40, Height: 60 mm, Diameter: 42 mm